5-Chlorbenzoxazol 95%
Lieferant: Thermo Scientific
|
Achtung
|
H27083.06
H27083.06EA
65.7
EUR
H27083.06
5-Chlorbenzoxazol 95%
5-Chlorobenzoxazole
Purity: 95%
CAS No.: 17200-29-2
Molecular Formula: C7H4ClNO
Formula Weight: 153.57
Melting Point: 39-41°
MDL No.: MFCD00216939
Formel:
C₇H₄ClNO Molecular Weight: 153,57 g/mol Schmelzpunkt: 39…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216939 CAS-Nummer: 17200-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...