1-Methyl-1H-pyrazol-5-boronsäurepinakolester ≥98%

Lieferant: Thermo Scientific

Synonyme: 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4,4,5,5-Tetramethyl-2-(1-methylpyrazol-5-yl)-1,3,2-dioxaborolane, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Methylpyrazole-5-boronic acid Pinacol ester

H27094.MD
H27094.MDEA 0 EUR
H27094.MD
1-Methyl-1H-pyrazol-5-boronsäurepinakolester ≥98%
1-Methyl-1H-pyrazol-5-boronsäurepinakolester

Purity: 98% CAS No.: 847818-74-0 Molecular Formula: C10H17BN2O2 Formula Weight: 208.07 Melting Point: 58-61° MDL No.: MFCD05861380

Formel: C₁₀H₁₇BN₂O₂
Molecular Weight: 208,07 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05861380
CAS-Nummer: 847818-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR