4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester 95%
Lieferant: Thermo Fisher Scientific
H27097.03
H27097.03EA
102
EUR
H27097.03
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester 95%
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
Purity: 95%
CAS No.: 165881-36-7
Molecular Formula: C14H24BBrO2
Formula Weight: 315.06
Refractive Index: 1.4960
Optical Rotation: +18° (c=1 in chloroform)
MDL No.: MFCD09037500
Formel:
C₁₄H₂₄BBrO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037500 CAS-Nummer: 165881-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...