Boc-trans-4-hydroxy-L-proline 97%

Lieferant: Thermo Fisher Scientific

H27110.06 H27110.14
H27110.06EA 58.8 EUR
H27110.06 H27110.14
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline

Purity: 97% CAS No.: 13726-69-7 Molecular Formula: C10H17NO5 Formula Weight: 231.25 Melting Point: 123-127° MDL No.: MFCD00053370

Formel: C₁₀H₁₇NO₅
Schmelzpunkt: 123…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053370
CAS-Nummer: 13726-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR