5-Chlor-2-methoxyphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(5-Chloro-2-methoxyphenyl)boronic acid, 5-Chloro-2-methoxybenzeneboronic acid
H27127.14
H27127.14EA
167
EUR
H27127.14
5-Chlor-2-methoxyphenylboronsäure ≥97%
5-Chlor-2-methoxyphenylboronsäure
Purity: 97%
CAS No.: 89694-48-4
Molecular Formula: C7H8BClO3
Formula Weight: 186.40
Melting Point: 134-141°
MDL No.: MFCD01318966
Formel:
C₇H₈BClO₃ Molecular Weight: 186,40 g/mol Schmelzpunkt: 134…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318966 CAS-Nummer: 89694-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...