2,6-Dichlor-4-nitropyridin ≥97%

Lieferant: Thermo Scientific
Gefahr

H27131.06
H27131.06EA 111 EUR
H27131.06
2,6-Dichlor-4-nitropyridin ≥97%
2,6-Dichloro-4-nitropyridine

Purity: 97% CAS No.: 25194-01-8 Molecular Formula: C5H2Cl2N2O2 Formula Weight: 192.99 Melting Point: 94-98° MDL No.: MFCD05670545

Formel: C₅H₂Cl₂N₂O₂
Molecular Weight: 192,99 g/mol
Schmelzpunkt: 94…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05670545
CAS-Nummer: 25194-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR