2-Chlor-3-chinolincarbaldehyd 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27133.09
H27133.03
H27133.09EA
127
EUR
H27133.09
H27133.03
2-Chlor-3-chinolincarbaldehyd 98%
2-Chloroquinoline-3-carbaldehyde
Purity: 98%
CAS No.: 73568-25-9
Molecular Formula: C10H6ClNO
Formula Weight: 191.62
Melting Point: 148-150°
MDL No.: MFCD00130079
Formel:
C₁₀H₆ClNO Molecular Weight: 191.62 g/mol Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130079 CAS-Nummer: 73568-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...