2-Chlor-3-chinolincarbaldehyd 98%

Lieferant: Thermo Fisher Scientific
Achtung

H27133.09 H27133.03
H27133.09EA 127 EUR
H27133.09 H27133.03
2-Chlor-3-chinolincarbaldehyd 98%
2-Chloroquinoline-3-carbaldehyde

Purity: 98% CAS No.: 73568-25-9 Molecular Formula: C10H6ClNO Formula Weight: 191.62 Melting Point: 148-150° MDL No.: MFCD00130079

Formel: C₁₀H₆ClNO
Molecular Weight: 191.62 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00130079
CAS-Nummer: 73568-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR