Boc-trans-4-hydroxy-D-proline 99%
Lieferant: Thermo Scientific
Synonyme:
(2R,4S)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
H27155.03
H27155.03EA
198
EUR
H27155.03
Boc-trans-4-hydroxy-D-proline 99%
Boc-trans-4-hydroxy-D-proline
Purity: 99%
CAS No.: 147266-92-0
Molecular Formula: C10H17NO5
Formula Weight: 231.25
Storage Temperature: 2°C to 8°C
Melting Point: ca 123°
MDL No.: MFCD01861341
Formel:
C₁₀H₁₇NO₅ Molecular Weight: 231,25 g/mol Schmelzpunkt: ∼ 123 °C |
MDL:
MFCD01861341 CAS-Nummer: 147266-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...