2-Hydroxycyclohexanon dimer ≥90%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27199.14
H27199.06
H27199.14EA
174
EUR
H27199.14
H27199.06
2-Hydroxycyclohexanon dimer ≥90%
2-Hydroxycyclohexanon dimer
Purity: 90+%
CAS No.: 30282-14-5
Molecular Formula: C12H20O4
Formula Weight: 228.28
Flash Point: 79°(174°F)
Melting Point: 100-107°
Boiling Point: 83-86°/13mm
UN No.: 1325
MDL No.: MFCD00151157
Formel:
C₆H₁₀O₂ Siedepunkt: 83…86 °C (13 mmHg) Schmelzpunkt: 100…107 °C Flammpunkt: 79 °C (174 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151157 CAS-Nummer: 30282-14-5 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...