3-(Methylthio)benzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(Methylmercapto)benzoesäure

H27200.03 H27200.06 H27200.14
H27200.03EA 44 EUR
H27200.03 H27200.06 H27200.14
3-(Methylthio)benzoesäure ≥97%
3-(Methylthio)benzoesäure

Purity: 97% CAS No.: 825-99-0 Molecular Formula: C8H8O2S Formula Weight: 168.21 Melting Point: 126-132° MDL No.: MFCD00156993

Formel: C₈H₈O₂S
Molecular Weight: 168.22 g/mol
Schmelzpunkt: 126…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156993
CAS-Nummer: 825-99-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR