2,2,2-Trifluorethylformiat 95%

Lieferant: Thermo Fisher Scientific
Gefahr

H27212.06
H27212.06EA 94.2 EUR
H27212.06
2,2,2-Trifluorethylformiat 95%
2,2,2-Trifluorethylformiat

Purity: 95% CAS No.: 32042-38-9 Molecular Formula: HCO2CH2CF3 Formula Weight: 128.05 Flash Point: 7°(45°F) Refractive Index: 1.001 Boiling Point: 59-60° UN No.: 3272 MDL No.: MFCD06797733

Formel: C₃H₃F₃O₂
Molecular Weight: 128.05 g/mol
Siedepunkt: 59…60 °C
Dichte: 1.317
Flammpunkt: 7 °C (45 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD06797733
CAS-Nummer: 32042-38-9
UN: 3272
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR