2-(1-Naphthalin)ethanol 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(Naphthalen-1-yl)ethanol, 2-(1-Naphthyl)ethanol
H27218.09
H27218.09EA
34.6
EUR
H27218.09
2-(1-Naphthalin)ethanol 95%
2-(1-Naphthalin)ethanol
Purity: 95%
CAS No.: 773-99-9
Molecular Formula: C12H12O
Formula Weight: 172.23
Melting Point: 60-64°
Boiling Point: 186°/17mm
MDL No.: MFCD00004050
Formel:
C₁₂H₁₂O Molecular Weight: 172,23 g/mol Siedepunkt: 186 °C (17 mmHg) Schmelzpunkt: 60…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004050 CAS-Nummer: 773-99-9 EINECS: 212-260-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...