(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Boc-Cis-4-F-Pro-OH, N-Boc-cis-4-fluoro-L-proline, N-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid, (2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4S)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid
H27225.03
H27225.03EA
137
EUR
H27225.03
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97%
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Purity: 97%
CAS No.: 203866-13-1
Molecular Formula: C10H16FNO4
Formula Weight: 233.24
Storage Temperature: 2°C to 8°C
MDL No.: MFCD04973957
Formel:
C₁₀H₁₆FNO₄ Molecular Weight: 233.24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04973957 CAS-Nummer: 203866-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...