6-Brom-2-pyridinboronsäurepinakolester 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H27245.03
H27245.03EA 303 EUR
H27245.03
6-Brom-2-pyridinboronsäurepinakolester 95%
6-Brom-2-pyridinboronsäurepinakolester

Purity: 95% CAS No.: 651358-83-7 Molecular Formula: C11H15BBrNO2 Formula Weight: 283.96 Melting Point: 125-127° MDL No.: MFCD06796177

Formel: C₁₁H₁₅BBrNO₂
Molecular Weight: 283,96 g/mol
Schmelzpunkt: 125…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06796177
CAS-Nummer: 651358-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR