4-Cyano-3-iodopyridine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27247.03
H27247.03EA
357
EUR
H27247.03
4-Cyano-3-iodopyridine 98%
4-Cyano-3-iodopyridine
Purity: 98%
CAS No.: 10386-28-4
Molecular Formula: C6H3IN2
Formula Weight: 230.01
Melting Point: 119-122°
UN No.: UN3439
MDL No.: MFCD08729285
Formel:
C₆H₃IN₂ Schmelzpunkt: 119…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08729285 CAS-Nummer: 10386-28-4 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...