5-Brom-2'-deoxyuridin (Broxuridin) ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 59-14-3
Molecular Formula: C9H11BrN2O5
Formula Weight: 307.10
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 30395
Merck Reference: 14,1452
Optical Rotation: +31° (c=1 in 0.1N HCl)
MDL No.: MFCD00006529
Formel:
C₉H₁₁BrN₂O₅ Schmelzpunkt: ∼ 190 °C (Zersetzung) |
MDL:
MFCD00006529 CAS-Nummer: 59-14-3 EINECS: 200-415-9 Merck Index: 14,1452 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...