Tetrabutylammoniumdihydrogentrifluorid tech. 90%, technische Qualität
Lieferant: Thermo Scientific
|
Gefahr
|
H27266.06
H27266.06EA
141
EUR
H27266.06
Tetrabutylammoniumdihydrogentrifluorid tech. 90%, technische Qualität
Tetrabutylammoniumdihydrogentrifluorid
Purity: 90%
CAS No.: 99337-56-1
Molecular Formula: [CH3(CH2)3)]4N(H2F3)
Formula Weight: 301.48
UN No.: 3265
MDL No.: MFCD00145365
Formel:
C₁₆H₃₈F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145365 CAS-Nummer: 99337-56-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...