1-(5-Iodopyridin-2-yl)piperazine 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H27289.03
H27289.03EA
128
EUR
H27289.03
1-(5-Iodopyridin-2-yl)piperazine 95%
1-(5-Iodopyridin-2-yl)piperazine
Purity: 95%
CAS No.: 219635-89-9
Molecular Formula: C9H12IN3
Formula Weight: 289.12
Melting Point: ca 135° dec.
MDL No.: MFCD04039867
Formel:
C₉H₁₂IN₃ Schmelzpunkt: ∼ 135 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039867 CAS-Nummer: 219635-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...