4-Oxocyclohexancarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H27294.03
H27294.03EA
111
EUR
H27294.03
4-Oxocyclohexancarbonsäure ≥98%
4-Oxocyclohexanecarboxylic acid
Purity: 98%
CAS No.: 874-61-3
Molecular Formula: C7H10O3
Formula Weight: 142.16
Melting Point: 65-71°
MDL No.: MFCD00102035
Formel:
C₇H₁₀O₃ Molecular Weight: 142,16 g/mol Schmelzpunkt: 65…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00102035 CAS-Nummer: 874-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...