2-Chloro-4-(trifluoromethyl)nicotinic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

H27312.03
H27312.03EA 109 EUR
H27312.03
2-Chloro-4-(trifluoromethyl)nicotinic acid 97%
2-Chloro-4-(trifluoromethyl)nicotinic acid

Purity: 97% CAS No.: 590371-81-6 Molecular Formula: C7H3ClF3NO2 Formula Weight: 225.56 Melting Point: 158-159° MDL No.: MFCD08458125

Formel: C₇H₃ClF₃NO₂
Schmelzpunkt: 158…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458125
CAS-Nummer: 590371-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR