(2,5-Dimethoxyphenyl)essigsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27328.06
H27328.14
H27328.06EA
55.1
EUR
H27328.06
H27328.14
(2,5-Dimethoxyphenyl)essigsäure ≥99%
(2,5-Dimethoxyphenyl)essigsäure
Purity: 99%
CAS No.: 1758-25-4
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 123-125°
MDL No.: MFCD00004322
Formel:
(C₆H₃(OCH₃)₂)CH₂COOH Molecular Weight: 196.2 g/mol Siedepunkt: 327 Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004322 CAS-Nummer: 1758-25-4 EINECS: 217-151-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...