1-Butyl-3-methylimidazoliummethansulfonat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-n-Butyl-3-methylimidazolium methanesulphonate

H27336.06 H27336.18
H27336.06EA 57.7 EUR
H27336.06 H27336.18
1-Butyl-3-methylimidazoliummethansulfonat ≥99%
1-Butyl-3-methylimidazolium methanesulphonate

Purity: 99% CAS No.: 342789-81-5 Molecular Formula: C9H18N2O3S Formula Weight: 234.32 Flash Point: 119°(246°F) Melting Point: 75-80° UN No.: 3261 MDL No.: MFCD06798173

Formel: C₉H₁₈N₂O₃S
Molecular Weight: 234.32 g/mol
Schmelzpunkt: 75…80 °C
Flammpunkt: 119 °C (246 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD06798173
CAS-Nummer: 342789-81-5
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR