1-Methylimidazol-5-carbaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27338.03
H27338.03EA
117
EUR
H27338.03
1-Methylimidazol-5-carbaldehyd ≥97%
1-Methylimidazol-5-carbaldehyd
Purity: 97%
CAS No.: 39021-62-0
Molecular Formula: C5H6N2O
Formula Weight: 110.12
Flash Point:
Formel:
C₅H₆N₂O Schmelzpunkt: 52…57 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00030439 CAS-Nummer: 39021-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...