9,9-Dimethylxanthene ≥98%
Lieferant: Thermo Scientific
Synonyme:
9,9-Dimethyl-9H-xanthene
H27340.06
H27340.06EA
129
EUR
H27340.06
9,9-Dimethylxanthene ≥98%
9,9-Dimethylxanthene
Purity: 98+%
CAS No.: 19814-75-6
Molecular Formula: C15H14O
Formula Weight: 210.28
Flash Point:
Formel:
C₁₅H₁₄O Molecular Weight: 210,28 g/mol Siedepunkt: 114…115 °C (0,6 mmHg) Schmelzpunkt: 40…43 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134434 CAS-Nummer: 19814-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...