N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥95%

Lieferant: Thermo Scientific

Synonyme: Fmoc-Cys(Trt)-OH, N-Fmoc-S-trityl-L-cysteine

H27363.06
H27363.06EA 42.1 EUR
H27363.06
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine ≥95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine

Purity: 95% CAS No.: 103213-32-7 Molecular Formula: C37H31NO4S Formula Weight: 585.71 Storage Temperature: 2°C to 8°C Melting Point: 171-173° Optical Rotation: +17° (c=1 in THF) MDL No.: MFCD00038538

Formel: C₃₇H₃₁NO₄S
Molecular Weight: 585,72 g/mol
Schmelzpunkt: 171…173 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00038538
CAS-Nummer: 103213-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR