Kalium(4-methoxyphenyl)trifluoroborat 95%

Lieferant: Thermo Fisher Scientific
Achtung

H27384.06 H27384.03
H27384.06EA 184 EUR
H27384.06 H27384.03
Kalium(4-methoxyphenyl)trifluoroborat 95%
Kalium(4-methoxyphenyl)trifluoroborat

Purity: 95% CAS No.: 192863-36-8 Molecular Formula: C7H7BF3KO Formula Weight: 214.04 Melting Point: 300° MDL No.: MFCD04112717

Formel: C₇H₇BF₃KO
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04112717
CAS-Nummer: 192863-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR