3-Chlor-p-kresol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Chlor-4-hydroxytoluol, 3-Chlor-4-methylphenol, 1-Chlor-3-hydroxy-6-methylbenzol
H27385.06
H27385.06EA
74.5
EUR
H27385.06
3-Chlor-p-kresol ≥97%
3-Chlor-p-kresol
Purity: 97%
CAS No.: 615-62-3
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Melting Point: 51-53°
Boiling Point: 228°
UN No.: 3437
MDL No.: MFCD00060319
Formel:
C₇H₇ClO Molecular Weight: 142,58 g/mol Siedepunkt: 228 °C Schmelzpunkt: 51…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060319 CAS-Nummer: 615-62-3 EINECS: 210-439-1 UN: 3437 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...