4-Benzyloxy-6-chloropyrimidine 95%

Lieferant: Thermo Fisher Scientific
Achtung

H27390.MD H27390.03
H27390.MDEA 143 EUR
H27390.MD H27390.03
4-Benzyloxy-6-chloropyrimidine 95%
4-Benzyloxy-6-chloropyrimidine

Purity: 95% CAS No.: 405930-65-6 Molecular Formula: C11H9ClN2O Formula Weight: 220.66 MDL No.: MFCD06798234

Formel: C₁₁H₉ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06798234
CAS-Nummer: 405930-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR