trans-4-Hydroxy-D-proline 97%
Lieferant: Thermo Fisher Scientific
H27421.MD
H27421.03
H27421.MDEA
89.6
EUR
H27421.MD
H27421.03
trans-4-Hydroxy-D-proline 97%
trans-4-Hydroxy-D-proline
Purity: 97%
CAS No.: 3398-22-9
Molecular Formula: C5H9NO3
Formula Weight: 131.13
Melting Point: ca 260° dec.
MDL No.: MFCD00065608
Formel:
C₅H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065608 CAS-Nummer: 3398-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...