6-Chlor-2-pyridinboronsäurepinakolester tech. 90%, technische Qualität

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Chloropyridine-2-boronic acid pinacol ester

H27426.03
H27426.03EA 404 EUR
H27426.03
6-Chlor-2-pyridinboronsäurepinakolester tech. 90%, technische Qualität
6-Chlor-2-pyridinboronsäurepinakolester

Purity: 90% CAS No.: 652148-92-0 Molecular Formula: C11H15BClNO2 Formula Weight: 239.51 Storage Temperature: -30°C to -10°C Melting Point: 107-110° MDL No.: MFCD06798259

Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239,51 g/mol
Schmelzpunkt: 107…110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD06798259
CAS-Nummer: 652148-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR