6-(Boc-methylamino)-3-pyridinboronsäurepinakolester 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, tert-Butyl methyl(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate

H27430.03 H27430.MD
H27430.03EA 253 EUR
H27430.03 H27430.MD
6-(Boc-methylamino)-3-pyridinboronsäurepinakolester 95%
6-(Boc-methylamino)-3-pyridinboronsäurepinakolester

Purity: 95% CAS No.: 1032758-87-4 Molecular Formula: C17H27BN2O4 Formula Weight: 334.22 Storage Temperature: 2°C to 8°C Melting Point: 112-119° MDL No.: MFCD09037484

Formel: C₁₇H₂₇BN₂O₄
Schmelzpunkt: 112…119 °C
MDL: MFCD09037484
CAS-Nummer: 1032758-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR