6-(Boc-methylamino)-3-pyridinboronsäurepinakolester 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, tert-Butyl methyl(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate
H27430.03
H27430.MD
H27430.03EA
253
EUR
H27430.03
H27430.MD
6-(Boc-methylamino)-3-pyridinboronsäurepinakolester 95%
6-(Boc-methylamino)-3-pyridinboronsäurepinakolester
Purity: 95%
CAS No.: 1032758-87-4
Molecular Formula: C17H27BN2O4
Formula Weight: 334.22
Storage Temperature: 2°C to 8°C
Melting Point: 112-119°
MDL No.: MFCD09037484
Formel:
C₁₇H₂₇BN₂O₄ Schmelzpunkt: 112…119 °C |
MDL:
MFCD09037484 CAS-Nummer: 1032758-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...