4-Benzyloxy-3-methoxybenzylalkohol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Methoxy-4-benzyloxybenzyl alcohol

H27443.06
H27443.06EA 81.2 EUR
H27443.06
4-Benzyloxy-3-methoxybenzylalkohol ≥98%
4-Benzyloxy-3-methoxybenzylalkohol

Purity: 98% CAS No.: 33693-48-0 Molecular Formula: C15H16O3 Formula Weight: 244.30 Melting Point: 71-73° MDL No.: MFCD00004640

Formel: C₁₅H₁₆O₃
Molecular Weight: 244,29 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004640
CAS-Nummer: 33693-48-0
EINECS: 251-638-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR