2-(Benzylamino)-5-pyridinboronsäurepinakolester 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-(Benzylamino)pyridine-3-boronic acid pinacol ester, N-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

H27463.MD H27463.03
H27463.MDEA 73.3 EUR
H27463.MD H27463.03
2-(Benzylamino)-5-pyridinboronsäurepinakolester 95%
2-(Benzylamino)-5-pyridinboronsäurepinakolester

Purity: 95% CAS No.: 1073354-27-4 Molecular Formula: C18H23BN2O2 Formula Weight: 310.20 Melting Point: 135-138° MDL No.: MFCD06798270

Formel: C₁₈H₂₃BN₂O₂
Molecular Weight: 310.20 g/mol
Schmelzpunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798270
CAS-Nummer: 1073354-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR