D(-)-Isoleucin 98

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3R)-(-)-α-Amino-β-methylvaleriansäure, (2R,3R)-(-)-2-Amino-3-methylpentansäure, (2R,3R)-(-)-Isoleucin, H-D-Ile-OH

H27488.03 H27488.MD
H27488.03EA 455 EUR
H27488.03 H27488.MD
D(-)-Isoleucin 98
D(-)-Isoleucin

Purity: 98% CAS No.: 319-78-8 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 265° subl. Optical Rotation: -40° (c=5 in 5N HCl)

Formel: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Molecular Weight: 131.17 g/mol
Siedepunkt: 235
Schmelzpunkt: 265 °C (Subl.)
Dichte: 1.224 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064221
CAS-Nummer: 319-78-8
EINECS: 206-269-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR