Diphenyl(o-tolyl)phosphine
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27503.03
H27503.03EA
34.4
EUR
H27503.03
Diphenyl(o-tolyl)phosphine
Diphenyl(o-tolyl)phosphine
CAS No.: 5931-53-3
Molecular Formula: C19H17P
Formula Weight: 276.32
Melting Point: 67-70°
MDL No.: MFCD00008515
Formel:
C₁₉H₁₇P Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008515 CAS-Nummer: 5931-53-3 EINECS: 227-675-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...