2,6-Difluor-3-pyridinboronsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,6-Difluoropyridine-3-boronic acid
H27509.03
H27509.03EA
174
EUR
H27509.03
2,6-Difluor-3-pyridinboronsäure 95%
2,6-Difluoro-3-pyridineboronic acid
Purity: 95%
CAS No.: 136466-94-9
Molecular Formula: C5H4BF2NO2
Formula Weight: 158.90
MDL No.: MFCD03788245
Formel:
C₅H₄BF₂NO₂ Molecular Weight: 158,90 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788245 CAS-Nummer: 136466-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...