(S)-(-)-1-Boc-3-methylpiperazin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-tert-Butyl 3-methylpiperazine-1-carboxylate, (3S)-3-Methylpiperazine, N1-BOC protected
H27534.06
H27534.03
H27534.06EA
216
EUR
H27534.06
H27534.03
(S)-(-)-1-Boc-3-methylpiperazin ≥98%
(S)-(-)-1-Boc-3-methylpiperazin
Purity: 98%
CAS No.: 147081-29-6
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Melting Point: 38-41°
Optical Rotation: -16° (c=1 in dioxane)
Formel:
C₁₀H₂₀N₂O₂ Schmelzpunkt: 38…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683204 CAS-Nummer: 147081-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...