3-Amino-2-methylpyridin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Methylpyridin-3-amin, 3-Amino-2-picolin, 2-Methyl-3-pyridylamin

H27540.03
H27540.03EA 86.5 EUR
H27540.03
3-Amino-2-methylpyridin 97%
3-Amino-2-methylpyridin

Purity: 97% CAS No.: 3430-10-2 Molecular Formula: C6H8N2 Formula Weight: 108.14 Odour: Characteristic Melting Point: 114-119° UN No.: 2811 MDL No.: MFCD03788195

Formel: C₆H₈N₂
Molecular Weight: 108,14 g/mol
Schmelzpunkt: 114…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788195
CAS-Nummer: 3430-10-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR