2-Chlor-4-hydroxypyridin 95%

Lieferant: Thermo Scientific
Achtung

H27549.03
H27549.03EA 143 EUR
H27549.03
2-Chlor-4-hydroxypyridin 95%
2-Chlor-4-hydroxypyridin

Purity: 95% CAS No.: 17368-12-6 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 169-172° MDL No.: MFCD01646108

Formel: C₅H₄ClNO
Molecular Weight: 129,55 g/mol
Schmelzpunkt: 169…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646108
CAS-Nummer: 17368-12-6
EINECS: 241-399-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR