2-Brom-4-methylbenzaldehyd 95%

Lieferant: Thermo Scientific
Achtung

H27558.03
H27558.03EA 71 EUR
H27558.03
2-Brom-4-methylbenzaldehyd 95%
2-Bromo-4-methylbenzaldehyde

Purity: 95% CAS No.: 824-54-4 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Melting Point: 33-37° MDL No.: MFCD04039890

Formel: C₈H₇BrO
Molecular Weight: 199,05 g/mol
Schmelzpunkt: 33…37 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD04039890
CAS-Nummer: 824-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR