4-Chlor-2-methoxybenzylalkohol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Chloro-2-methoxyphenyl)methanol

H27562.14
H27562.14EA 156 EUR
H27562.14
4-Chlor-2-methoxybenzylalkohol ≥97%
4-Chlor-2-methoxybenzylalkohol

Purity: 97% CAS No.: 55685-75-1 Molecular Formula: C8H9ClO2 Formula Weight: 172.61 Flash Point: 110°(230°F) Melting Point: 46-50° MDL No.: MFCD02683547

Formel: C₈H₉ClO₂
Molecular Weight: 172,61 g/mol
Schmelzpunkt: 46…50 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02683547
CAS-Nummer: 55685-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR