Methyl-5-methyl-2-furoat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 5-methylfuran-2-carboxylate

H27570.03
H27570.03EA 46.5 EUR
H27570.03
Methyl-5-methyl-2-furoat ≥97%
Methyl-5-methyl-2-furoat

Purity: 97% CAS No.: 2527-96-0 Molecular Formula: C7H8O3 Formula Weight: 140.14 Flash Point: 87°(189°F) Refractive Index: 1.493 Boiling Point: 255° MDL No.: MFCD04116374

Formel: C₇H₈O₃
Molecular Weight: 140.14 g/mol
Siedepunkt: 255 °C
Dichte: 1.114
Flammpunkt: 87 °C (189 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD04116374
CAS-Nummer: 2527-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR