Methyl-6-aminonicotinat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

H27575.06 H27575.03
H27575.06EA 84 EUR
H27575.06 H27575.03
Methyl-6-aminonicotinat 97%
Methyl-6-aminonicotinat

Purity: 97% CAS No.: 36052-24-1 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: 158-161° MDL No.: MFCD00797844

Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 158…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00797844
CAS-Nummer: 36052-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR