2,4-Dichlor-L-phenylalanin ≥98%

Lieferant: Thermo Scientific

Synonyme: H-Phe(2,4-DiCl)-OH

H27579.03
H27579.03EA 183 EUR
H27579.03
2,4-Dichlor-L-phenylalanin ≥98%
2,4-Dichlor-L-phenylalanin

Purity: 98% CAS No.: 111119-36-9 Molecular Formula: C9H9Cl2NO2 Formula Weight: 234.09 MDL No.: MFCD01860882

Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860882
CAS-Nummer: 111119-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR