2-Ethoxycyclohexanon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27613.06
H27613.06EA
48.4
EUR
H27613.06
2-Ethoxycyclohexanon ≥98%
2-Ethoxycyclohexanone
Purity: 98%
CAS No.: 33371-97-0
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 54°(129°F)
Refractive Index: 1.4510
Boiling Point: 85°/15mm
UN No.: 1224
MDL No.: MFCD00015377
Formel:
C₈H₁₄O₂ Siedepunkt: 85 °C (15 mmHg) Dichte: 1,002 Flammpunkt: 54 °C (129 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015377 CAS-Nummer: 33371-97-0 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...