2H-1,4-Benzoxazin-3(4H)-on 99%

Lieferant: Thermo Fisher Scientific
Achtung

H27622.14 H27622.06
H27622.14EA 77.1 EUR
H27622.14 H27622.06
2H-1,4-Benzoxazin-3(4H)-on 99%
2H-1,4-Benzoxazin-3(4H)-one

Purity: 99% CAS No.: 5466-88-6 Molecular Formula: C8H7NO2 Formula Weight: 149.15 Melting Point: 173-175° MDL No.: MFCD00158536

Formel: C₈H₇NO₂
Molecular Weight: 149.15 g/mol
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00158536
CAS-Nummer: 5466-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR