1-(1-Methylpiperidin-4-yl)piperazine 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H27624.06
H27624.03
H27624.06EA
164
EUR
H27624.06
H27624.03
1-(1-Methylpiperidin-4-yl)piperazine 98%
1-(1-Methylpiperidin-4-yl)piperazine
Purity: 98%
CAS No.: 23995-88-2
Molecular Formula: C10H21N3
Formula Weight: 183.30
Refractive Index: 1.508
Melting Point: 29-32°
UN No.: UN3259
MDL No.: MFCD01075186
Formel:
C₁₀H₂₁N₃ Schmelzpunkt: 29…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075186 CAS-Nummer: 23995-88-2 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...