(R)-(+)-Indolin-2-carbonsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H27626.03
H27626.03EA 95 EUR
H27626.03
(R)-(+)-Indolin-2-carbonsäure ≥97%
(R)-(+)-Indolin-2-carbonsäure

Purity: 97% CAS No.: 98167-06-7 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: ca 177° dec. MDL No.: MFCD00792496

Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Schmelzpunkt: ∼ 177 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00792496
CAS-Nummer: 98167-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR